missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Porcine IFN-gamma Recombinant Protein

Brand:  R&D Systems™ 985-PI-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486484

  • £352.00 / 50µg


Estimated Shipment
28-06-2018
Add to basket

Description

Description

Source: E. coli-derived, Ser21-Lys166
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 396991
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
17.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Porcine
Yes
396991
IFN-gamma
50μg
Unlabeled
Videos