missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Porcine GM-CSF Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 397208 |
Molecular Weight (g/mol) | 14.4 kDa |
Name | GM-CSF |
Quantity | 10μg |
Description
The Recombinant Porcine GM-CSF Protein is derived from E. coli. The Recombinant Porcine GM-CSF Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
14.4 kDa | |
10μg | |
E. coli-derived porcine GM-CSF protein Ala18-Lys144 |
397208 | |
GM-CSF | |
Yes |