Learn More
R&D Systems™ PARP Enzyme High Specific Activity
Purified from E. coli containing a recombinant plasmid harboring the human PARP gene
Brand:  R&D Systems™ 4668-100-01
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13445024
  • £318.13 / 100µL
Estimated Shipment
27-06-2019
Description
Description
PARP [Poly(ADP-ribose) Polymerase], also known as ADPRT and PPOL, is a 118kDa enzyme that uses NAD as a substrate to catalyze the covalent transfer of ADP-ribose to a variety of nuclear protein acceptors. ADP ribosyltransferase is required for cellular repair, and PARP expression is induced by single-strand breaks in DNA. PARP is proteolytically cleaved by Caspase-3 into two fragments of 89 and 24kDa in one of the hallmark events of apoptosis.
  • One unit of PARP incorporates 10femtomoles of NAD onto immobilized histones in 30 minutes
  • High specific activity PARP is provided at 10 units/μL
Specifications
Specifications
Poly [ADP-ribose] Polymerase Enzyme High Specific Activity
100μL
Store at -20° C in a manual defrost freezer. For long-term storage, freeze in working aliquots at -80° C. Avoid freeze-thaw cycles.
Videos