missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse VEGF R2/KDR/Flk-1 Fc Chimera Recombinant Protein

Brand:  R&D Systems™ 443-KD-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464764

  • £351.13 / 50µg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala20-Glu726
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 110 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16542
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
110 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
16542
VEGF R2/KDR/Flk-1 Fc Chimera
50μg
Disulfide-linked homodimer
Yes
Videos