missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse VEGF-D Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
POA
Specifications
Gene ID (Entrez) | 14205 |
---|---|
Molecular Weight (g/mol) | 13 kDa |
Name | VEGF-D |
Quantity | 25μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse VEGF-D protein Phe98-Ser206,with a C-terminal 6-His tag |
Description
The Recombinant Mouse VEGF-D Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse VEGF-D Protein has been validated for the following applications: Bioactivity, Binding Activity.Specifications
14205 | |
VEGF-D | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse VEGF-D protein Phe98-Ser206,with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
13 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |