missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse VEGF 164 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 22339 |
---|---|
Molecular Weight (g/mol) | 19.4 kDa (monomer) |
Name | VEGF 164 |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse VEGF protein Ala27-Arg190 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Description
The Recombinant Mouse VEGF 164 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse VEGF 164 Protein has been validated for the following applications: Bioactivity.Specifications
22339 | |
VEGF 164 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
19.4 kDa (monomer) | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse VEGF protein Ala27-Arg190 | |
Disulfide-linked homodimer | |
Recombinant |