missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse VEGF 164 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 493-MV-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445194

  • £595.17 / 25µg

Estimated Shipment date
from Supplier 24-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse VEGF 164 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse VEGF 164 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
19.4 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Disulfide-linked homodimer
Yes
22339
VEGF 164
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents