missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse ULBP-1 /MULT-1 Recombinant Protein

Brand:  R&D Systems™ 2588-MU-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461524

  • £316.81 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Pro26-Thr211, with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 77777
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
22 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
77777
ULBP-1 /MULT-1
50μg
Unlabeled
Videos