missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TrkB Recombinant Protein

Brand:  R&D Systems™ 1494-TB-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459363

  • £298.33 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Cys32-His429, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 45.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 18212
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
45.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
18212
TrkB
25μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos