missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TNF RII/TNFRSF1B Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 426-R2-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13444464

  • £445.48 / 50µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse sTNF RII/TNFRSF1B Protein is derived from E. coli. The Recombinant Mouse sTNF RII/TNFRSF1B Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
25 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived mouse TNF RII/TNFRSF1B protein Val23-Gly258
Unconjugated
21938
TNF RII/TNFRSF1B
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents