Learn More
R&D Systems™ Mouse TNF-alpha ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Brand:  R&D Systems™ EL410
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13435115
  • £443.94 / 1 set
Estimated Shipment date
from Supplier 26-02-2020
Add to basket
Specifications
Specifications
Diagnostics
Mouse TNF-alpha ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
One 96-well PVDF-backed plate coated w/pAb for mouse TNF-a, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control
Whole Cells
One 96-well plate