missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse TNF-alpha ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
POA
Specifications
Product Type | Mouse TNF-alpha ELISpot Kit |
---|---|
For Use With (Application) | Diagnostics |
Sample Type | Whole Cells |
Specifications
Mouse TNF-alpha ELISpot Kit | |
Diagnostics | |
Whole Cells |