missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse TNF-alpha aa 84-235 Recombinant Protein

Brand:  R&D Systems™ 410-TRNC-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464164

  • £500.52 / 50µg


Estimated Shipment
08-01-2019
Add to basket

Description

Description

Source: E. coli-derived, Ser84-Leu235
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21926
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
17 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
21926
TNF-alpha aa 84-235
50μg
Unlabeled
Videos