missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse SOST/Sclerostin Recombinant Protein

Brand:  R&D Systems™ 1589-ST-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459743

  • £368.29 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Gln24-Tyr211 with an N-terminal 7-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 74499
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
22 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
74499
SOST/Sclerostin
25μg
Unlabeled
Videos