missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse SCGF/CLEC11a Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 20256 |
Molecular Weight (g/mol) | 35.1 kDa |
Name | SCGF/CLEC11a |
Quantity | 25μg |
Description
The Recombinant Mouse SCGF/CLEC11a Protein is derived from NS0. The Recombinant Mouse SCGF/CLEC11a Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
35.1 kDa | |
25μg | |
Mouse myeloma cell line,NS0-derived mouse SCGF/CLEC11a protein Ala22-Phe328,with a C-terminal 6-His tag |
20256 | |
SCGF/CLEC11a | |
Yes |