missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse SCGF/CLEC11a Recombinant Protein

Brand:  R&D Systems™ 3729-SC-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463504

  • £298.33 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala22-Phe328, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 35.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20256
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
35.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
20256
SCGF/CLEC11a
25μg
Unlabeled
Videos