missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Ret Fc Chimera Recombinant Protein

Brand:  R&D Systems™ 482-RT-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465124

  • £316.81 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Leu29-Arg637 (Phe174Ser)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 95 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 19713
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
95 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
19713
Ret Fc Chimera
100μg
Disulfide-linked homodimer
Yes
Videos