missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse R-Spondin 4 Recombinant Protein

Brand:  R&D Systems™ 4106-RS-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464154

  • £316.81 / 25µg


Estimated Shipment
12-04-2019
Add to basket

Description

Description

Measured by its ability to induce Topflash reporter activity in HEK293T human embryonic kidney cells. The ED 50 for this effect is 5-20 ng/mL in the presence of 10 ng/mL Recombinant Mouse Wnt-3a (Catalog # 1324-WN ).
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
23.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
228770
R-Spondin 4
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos