missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Prolactin Recombinant Protein

Brand:  R&D Systems™ 1445-PL-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459513

  • £217.80 / 50µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Leu32-Cys228, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 19109
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
22.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
19109
Prolactin
50μg
Unlabeled
Videos