missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Prolactin Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1445-PL-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13469513

  • £257.52 / 50µg

Estimated Shipment
17-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse Prolactin Protein is derived from E. coli. The Recombinant Mouse Prolactin Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
22.6 kDa
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
E. coli-derived mouse Prolactin protein Leu32-Cys228,with an N-terminal Met
Unconjugated
19109
Prolactin
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents