missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse P-Selectin/CD62P Fc Chimera Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 737-PS-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13456154

  • £244.02 / 50µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse P-Selectin/CD62P Fc Chimera Protein is derived from NS0. The Recombinant Mouse P-Selectin/CD62P Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
99.1 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
NS0
Disulfide-linked homodimer
Yes
20344
P-Selectin/CD62P Fc Chimera
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents