missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse P-Selectin/CD62P Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
---|---|
Gene ID (Entrez) | 20344 |
Molecular Weight (g/mol) | 99.1 kDa (monomer) |
Name | P-Selectin/CD62P Fc Chimera |
Source | Mouse myeloma cell line,NS0-derived mouse P-Selectin/CD62P protein, (N-terminus) Mouse P-Selectin (Trp42-Ala709) Accession &Num; Q01102 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Description
The Recombinant Mouse P-Selectin/CD62P Fc Chimera Protein is derived from NS0. The Recombinant Mouse P-Selectin/CD62P Fc Chimera Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
99.1 kDa (monomer) | |
Mouse myeloma cell line,NS0-derived mouse P-Selectin/CD62P protein, (N-terminus) Mouse P-Selectin (Trp42-Ala709) Accession &Num; Q01102 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
20344 | |
P-Selectin/CD62P Fc Chimera | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |