missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse OX40 Ligand/TNFSF4 Recombinant Protein

Brand:  R&D Systems™ 1236-OX-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458933

  • £364.33 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Gln49-Leu198, with an N-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 22164
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
18 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
22164
OX40 Ligand/TNFSF4
25μg
Unlabeled
Videos