missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Osteopontin/OPN Recombinant Protein

Brand:  R&D Systems™ 441-OP-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464724

  • £256.00 / 50µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Leu17-Asn294 (Glu99Gly), with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 31.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20750
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
31.5 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
20750
Osteopontin/OPN
50μg
Unlabeled
Videos