missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Netrin-4 Recombinant Protein

Brand:  R&D Systems™ 1132-N4-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458633

  • £316.81 / 25µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Leu19-Val628 & Ala26-Val628, both with a C-terminal 10-His tag
Purity: 80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 69 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 57764
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
69 kDa
80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
57764
Netrin-4
25μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos