missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Netrin-1 Recombinant Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 1109-N1-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468543

  • £421.89 / 25µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse Netrin-1 Protein is derived from NS0. The Recombinant Mouse Netrin-1 Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
68.2 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line, NS0-derived Mouse Netrin-1 (Val22-Ala603) Accession # AAC52971 IEGR GGGSGGGSGGGS 10-His tag
Unlabeled
18208
Netrin-1
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents