missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse MSP/MST1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 15235 |
---|---|
Molecular Weight (g/mol) | 53.7 kDa ( alpha-chain) & 25 kDa (beta-chain) |
Name | MSP/MST1 |
Quantity | 25μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse MSP/MST1 protein Met1-Glu716 (Cys677Ala) |
Description
The Recombinant Mouse MSP/MST1 Protein is derived from CHO. The Recombinant Mouse MSP/MST1 Protein has been validated for the following applications: Bioactivity.Specifications
15235 | |
MSP/MST1 | |
Chinese Hamster Ovary cell line,CHO-derived mouse MSP/MST1 protein Met1-Glu716 (Cys677Ala) | |
Disulfide-linked heterodimer | |
Recombinant |
53.7 kDa ( alpha-chain) & 25 kDa (beta-chain) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |