missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse MIF Recombinant Protein

Brand:  R&D Systems™ 1978-MF-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460344

  • £237.61 / 25µg


Estimated Shipment
03-01-2019
Add to basket

Description

Description

Source: E. coli-derived, Pro2-Ala115
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 17319
Regulatory Status: RUO

Specifications

Specifications

Reconstitute at 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
17319
MIF
25μg
Unlabeled
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
12.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
Videos