missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Meteorin Recombinant Protein

Brand:  R&D Systems™ 3475-MN-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463054

  • £298.33 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Gly22-Asp291 with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 30 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 70083
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
30 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
70083
Meteorin
25μg
Unlabeled
Videos