missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Meteorin Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 70083 |
Molecular Weight (g/mol) | 30 kDa |
Name | Meteorin |
Quantity | 25μg |
Description
The Recombinant Mouse Meteorin Protein is derived from NS0. The Recombinant Mouse Meteorin Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
30 kDa | |
25μg | |
Mouse myeloma cell line,NS0-derived mouse Meteorin protein Gly22-Asp291 with a C-terminal 6-His tag |
70083 | |
Meteorin | |
Yes |