missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse MBL-2 Recombinant Protein

Brand:  R&D Systems™ 2208-MB-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460804

  • £364.33 / 50µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Glu19-Asp244
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 24 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 17195
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
24 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
17195
MBL-2
50μg
Oligomer
Yes
Videos