missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Matrilin-2 Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Molecular Weight (g/mol) | 104.9 kDa |
Name | Matrilin-2 |
Quantity | 50μg |
Recombinant | Yes |
Description
The Recombinant Mouse Matrilin-2 Protein is derived from NS0. The Recombinant Mouse Matrilin-2 Protein has been validated for the following applications: Binding Activity.Specifications
Unconjugated | |
Matrilin-2 | |
Yes |
104.9 kDa | |
50μg | |
Mouse myeloma cell line,NS0-derived mouse Matrilin-2 protein Arg24-Arg956 & Met40-Arg956,both with a C-terminal 6-His tag |