missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse M-CSF Recombinant Protein

Brand:  R&D Systems™ 416-ML-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464274

  • £244.21 / 10µg


Estimated Shipment
18-12-2018
Add to basket

Description

Description

Source: E. coli-derived, Lys33-Glu262, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 26 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 12977
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
26 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
12977
M-CSF
10μg
Disulfide-linked homodimer
Yes
Videos