missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Leptin R Fc Chimera Recombinant Protein, Ala20-Gly839

Brand:  R&D Systems™ 497-LR-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465214

  • £332.65 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala20-Gly839
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 120 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS.
Entrez Gene ID: 16847
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS with BSA as a carrier protein.
120 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
16847
Leptin R Fc Chimera
100μg
Disulfide-linked homodimer
Yes
Videos