missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse KGF/FGF-7 Recombinant Protein

Brand:  R&D Systems™ 5028-KG-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465244

  • £356.00 / 25µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Source: E. coli-derived, Cys32-Thr194
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
KGF/FGF-7
25μg
Unlabeled
18.7 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
Videos