missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-6 R alpha Recombinant Protein

Brand:  R&D Systems™ 1830-SR-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460094

  • £245.53 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Leu20-Glu357, with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 39 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16194
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
39 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
16194
IL-6 R alpha
25μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos