missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-36Ra/IL-1F5 Recombinant Protein

Brand:  R&D Systems™ 2714-ML-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461784

  • £245.53 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Val2-Asp156
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 54450
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
54450
IL-36Ra/IL-1F5
25μg
Unlabeled
Videos