missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-36 gamma/IL-1F9 (aa 13-164) Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 215257 |
Molecular Weight (g/mol) | 17.3 kDa |
Name | IL-36 gamma/IL-1F9 (aa 13-164) |
Quantity | 10μg |
Description
The Recombinant Mouse IL-36 gamma/IL-1F9 (aa 13-164) Protein is derived from E. coli. The Recombinant Mouse IL-36 gamma/IL-1F9 (aa 13-164) Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
17.3 kDa | |
10μg | |
E. coli-derived mouse IL-36 gamma/IL-1F9 protein Gly13-Ser164 |
215257 | |
IL-36 gamma/IL-1F9 (aa 13-164) | |
Yes |