missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-36 alpha/IL-1F6 Recombinant Protein

Brand:  R&D Systems™ 2297-ML-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460964

  • £298.33 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Met1-His160
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 54448
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
21 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
54448
IL-36 alpha/IL-1F6
25μg
Unlabeled
Videos