missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-33 Recombinant Protein

Brand:  R&D Systems™ 3626-ML-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463314

  • £341.89 / 10µg


Estimated Shipment
29-03-2019
Add to basket

Description

Description

Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. The ED 50 for this effect is 0.0125-0.05 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
18 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
77125
IL-33
10μg
12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution.
Yes
Videos