missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-3 Recombinant Protein

For use in cell or tissue culture applications.

Brand:  R&D Systems™ 403-ML-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464004

  • £843.48 / 50µg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

  • Source: E. coli-derived, Asp33-Cys166, with an N-terminal Met
  • Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: 15 kDa
  • Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
  • Entrez Gene ID: 16187
  • Regulatory Status: RUO
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
15 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
16187
IL-3
50μg
Unlabeled
Videos