missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-20 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 58181 |
---|---|
Name | IL-20 |
Quantity | 25μg |
Source | E. coli-derived mouse IL-20 protein Leu25-Leu176,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. |
Description
The Recombinant Mouse IL-20 Protein is derived from E. coli. The Recombinant Mouse IL-20 Protein has been validated for the following applications: Bioactivity.Specifications
58181 | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. | |
Recombinant |
IL-20 | |
E. coli-derived mouse IL-20 protein Leu25-Leu176,with an N-terminal Met | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |