missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-2 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 402-ML-020

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13453984

  • £192.57 / 20µg

Estimated Shipment date
from Supplier 27-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse IL-2 Protein is derived from E. coli. The Recombinant Mouse IL-2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80 with BSA as a carrier protein.
17.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Reconstitute at 100-200μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
16183
IL-2
20μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents