missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-2 Recombinant Protein

Brand:  R&D Systems™ 402-ML-020/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463984

  • £167.64 / 20µg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. Gearing, A.J.H. and C.B. Bird (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 295. The ED 50 for this effect is 0.1-0.4 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80.
17.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Reconstitute at 100-200μg/mL in sterile deionized water.
16183
IL-2
20μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos