missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-2 ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
POA
Specifications
Antigen | IL-2 |
---|---|
Cross Reactivity | Mouse |
Product Type | Mouse IL-2 ELISpot Kit |
Includes | Mouse IL-2 Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Mouse IL-2 Positive Control |
Sample Type | Whole cells |
Specifications
IL-2 | |
Mouse IL-2 ELISpot Kit | |
Whole cells |
Mouse | |
Mouse IL-2 Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Mouse IL-2 Positive Control |