Learn More
R&D Systems™ Mouse IL-2 ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Brand:  R&D Systems™ XEL402
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13453296
  • £2529.87 / Pack of 6
Estimated Shipment date
from Supplier 30-03-2020
Add to basket
Specifications
Specifications
IL-2
Quantitative determination of the frequency of cells releasing mouse IL-2
Mouse IL-2 ELISpot Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Mouse
Mouse IL-2 Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Mouse IL-2 Positive Control
Whole cells
6 x 96-well plates