missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-18/IL-1F4, Carrier-Free Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ B004-2

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440455

  • £1100.74 / 200µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse IL-18/IL-1F4 Protein is derived from E. coli. The Recombinant Mouse IL-18/IL-1F4 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Bioactivity
Predicted MW: 18kDa
200μg
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Reconstitute in 2 mL of ice-cold distilled water on ice.
Lyophilized in PBS containing 1% sucrose.
>90%
E. coli-derived 36-192 aa
Yes
Videos