Learn More
R&D Systems™ Mouse IL-17 ELISpot Kit
highly sensitive, microplate-based assays for the detection of cytokine secreting cells
Brand:  R&D Systems™ EL421
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13465115
  • £390.73 / 1 set
Estimated Shipment
28-02-2019
Add to basket
Includes:
  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control
Description
Description
Product Features:
  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells
Specifications
Specifications
IL-17
Detection of cytokine secreting cells, quantitative sandwich ELISA
Mouse IL-17 ELISpot Kit
1 x 96-well plate
Mouse
Mouse IL-17/IL-17A Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Muse IL-17/IL-17A Positive Control
Whole cells
Videos