missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-13 Recombinant Protein

Brand:  R&D Systems™ 413-ML-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464224

  • £213.84 / 5µg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED 50 for this effect is 0.75-3 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
11.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
16163
IL-13
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos