missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-12 Recombinant Protein

Brand:  R&D Systems™ 419-ML-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464334

  • £282.49 / 10µg


Estimated Shipment
03-09-2018
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Met23-Ser335 (p40) & Arg23-Ala215 (p35)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 16159
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
IL-12
10μg
Disulfide-linked heterodimer
Yes
16159
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
Videos